Assay Details
| Gene Symbol: kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077151.1 | NP_001070619.1 | 4 - 5 | 697 | 115 | |
| GenBank mRNA | BC122284.1 | - | 4 - 5 | 697 | 115 | |
| BC157374.1 | - | 4 - 5 | 665 | 115 | ||
More Information
Set Membership: |
|

kctd13
Genomic Map