Assay Details
| Gene Symbol: gabpb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077152.1 | NP_001070620.1 | 4 - 5 | 514 | 117 | |
| XM_005158218.4 | XP_005158275.1 | 4 - 5 | 690 | 117 | ||
| GenBank mRNA | BC122444.1 | - | 4 - 5 | 514 | 117 | |
More Information
Set Membership: |
|

gabpb2b
Genomic Map