Assay Details
| Gene Symbol: proza | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327792.1 | NP_001314721.1 | 2 - 3 | 141 | 57 | |
| GenBank mRNA | BC124619.1 | - | 1 - 2 | 315 | 57 | |
More Information
Set Membership: |
|

proza
Genomic Map