Assay Details
| Gene Symbol: proza | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327792.1 | NP_001314721.1 | 5 - 6 | 449 | 67 | |
| GenBank mRNA | BC124619.1 | - | 4 - 5 | 623 | 67 | |
More Information
Set Membership: |
|

proza
Genomic Map