Assay Details
| Gene Symbol: spag1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077321.1 | NP_001070789.1 | 9 - 10 | 1078 | 60 | |
| GenBank mRNA | BC124658.1 | - | 9 - 10 | 1078 | 60 | |
More Information
Set Membership: |
|

spag1b
Genomic Map