Assay Details
| Gene Symbol: pros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077323.1 | NP_001070791.1 | 9 - 10 | 1285 | 83 | |
| GenBank mRNA | BC124790.1 | - | 9 - 10 | 1285 | 83 | |
| BC164563.1 | - | 9 - 10 | 1190 | 83 | ||
More Information
Set Membership: |
|

pros1
Genomic Map