Assay Details
| Gene Symbol: snx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077368.2 | NP_001070836.1 | 2 - 3 | 379 | 75 | |
| GenBank mRNA | BC124665.1 | - | 2 - 3 | 369 | 75 | |
More Information
Set Membership: |
|

snx21
Genomic Map