Assay Details
| Gene Symbol: scn1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077539.3 | NP_001071007.1 | 1 - 2 | 219 | 57 | |
| XM_005158211.4 | XP_005158268.1 | 1 - 2 | 1755 | 57 | ||
| GenBank mRNA | DQ489722.1 | - | 1 - 2 | 219 | 57 | |
| DQ489723.1 | - | 1 - 2 | 219 | 57 | ||
| DQ489724.1 | - | 1 - 2 | 219 | 57 | ||
| DQ489725.1 | - | 1 - 2 | 219 | 57 | ||
More Information
Set Membership: |
|

scn1ba
Genomic Map