Assay Details
| Gene Symbol: scn1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077539.3 | NP_001071007.1 | 3 - 4 | 627 | 82 | |
| XM_005158211.4 | XP_005158268.1 | 3 - 4 | 82 | |||
| XM_005158212.4 | XP_005158269.1 | 4 - 5 | 82 | |||
| XM_021466690.1 | XP_021322365.1 | 3 - 4 | 82 | |||
| GenBank mRNA | DQ489722.1 | - | 3 - 4 | 627 | 82 | |
| DQ489723.1 | - | 3 - 4 | 627 | 82 | ||
| DQ489725.1 | - | 3 - 4 | 627 | 82 | ||
More Information
Set Membership: |
|

scn1ba
Genomic Map