Assay Details
| Gene Symbol: mtfmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077542.2 | NP_001071010.2 | 7 - 8 | 1193 | 62 | |
| GenBank mRNA | BC142768.1 | - | 7 - 8 | 1194 | 62 | |
More Information
Set Membership: |
|

mtfmt
Genomic Map