Assay Details
| Gene Symbol: agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077551.1 | NP_001071019.1 | 4 - 5 | 895 | 81 | |
| GenBank mRNA | BC124326.1 | - | 4 - 5 | 895 | 81 | |
| BC164767.1 | - | 4 - 5 | 798 | 81 | ||
More Information
Set Membership: |
|

agmat
Genomic Map