Assay Details
| Gene Symbol: minpp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077557.1 | NP_001071025.1 | 1 - 2 | 689 | 102 | |
| GenBank mRNA | BC124216.1 | - | 1 - 2 | 689 | 102 | |
| BC164680.1 | - | 1 - 2 | 626 | 102 | ||
More Information
Set Membership: |
|

minpp1a
Genomic Map