Assay Details
| Gene Symbol: noxo1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077584.1 | NP_001071052.1 | 1 - 2 | 139 | 106 | |
| GenBank mRNA | AI959305.1 | - | 1 - 2 | 119 | 106 | |
| BC124172.1 | - | 1 - 2 | 139 | 106 | ||
More Information
Set Membership: |
|

noxo1a
Genomic Map