Assay Details
| Gene Symbol: smpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077615.1 | NP_001071083.1 | 3 - 4 | 1265 | 91 | |
| GenBank mRNA | BC124788.1 | - | 3 - 4 | 1265 | 91 | |
| BC165400.1 | - | 2 - 3 | 1130 | 91 | ||
More Information
Set Membership: |
|

smpd5
Genomic Map