Assay Details
| Gene Symbol: stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077616.1 | NP_001071084.1 | 2 - 3 | 1406 | 92 | |
| GenBank mRNA | BC125863.1 | - | 2 - 3 | 1406 | 92 | |
More Information
Set Membership: |
|

stk35
Genomic Map