Assay Details
| Gene Symbol: papss2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077767.1 | NP_001071235.1 | 4 - 5 | 571 | 99 | |
| GenBank mRNA | BC127397.1 | - | 4 - 5 | 571 | 99 | |
More Information
Set Membership: |
|

papss2a
Genomic Map