Assay Details
| Gene Symbol: papss2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077767.1 | NP_001071235.1 | 7 - 8 | 916 | 60 | |
| GenBank mRNA | BC127397.1 | - | 7 - 8 | 916 | 60 | |
More Information
Set Membership: |
|

papss2a
Genomic Map