Assay Details
| Gene Symbol: tsfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080035.1 | NP_001073504.1 | 4 - 5 | 484 | 88 | |
| GenBank mRNA | BC128869.1 | - | 4 - 5 | 484 | 88 | |
| BC152076.1 | - | 4 - 5 | 465 | 88 | ||
More Information
Set Membership: |
|

tsfm
Genomic Map