Assay Details
| Gene Symbol: tp53inp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080036.1 | NP_001073505.1 | 3 - 4 | 862 | 94 | |
| GenBank mRNA | BC127596.1 | - | 3 - 4 | 862 | 94 | |
| BC153523.1 | - | 2 - 3 | 640 | 94 | ||
More Information
Set Membership: |
|

tp53inp1
Genomic Map