Assay Details
| Gene Symbol: dalrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123326.1 | NP_001116798.1 | 3 - 4 | 972 | 76 | |
| GenBank mRNA | BC125925.1 | - | 3 - 4 | 915 | 76 | |
| BC154252.1 | - | 3 - 4 | 851 | 76 | ||
More Information
Set Membership: |
|

dalrd3
Genomic Map