Assay Details
| Gene Symbol: kdm2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081692.2 | NP_001075161.2 | 11 - 12 | 1520 | 108 | |
| XM_005160248.4 | XP_005160305.1 | 11 - 12 | 1478 | 108 | ||
| XM_009293357.3 | XP_009291632.1 | 11 - 12 | 2106 | 108 | ||
| XM_009293362.3 | XP_009291637.1 | 11 - 12 | 1477 | 108 | ||
| XM_009293365.3 | XP_009291640.1 | 11 - 12 | 1407 | 108 | ||
| GenBank mRNA | BC128874.1 | - | 2 - 3 | 316 | 108 | |
More Information
Set Membership: |
|

kdm2aa
Genomic Map