Assay Details
| Gene Symbol: kdm2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081692.2 | NP_001075161.2 | 18 - 19 | 3382 | 97 | |
| XM_005160248.4 | XP_005160305.1 | 18 - 19 | 3340 | 97 | ||
| XM_009293357.3 | XP_009291632.1 | 18 - 19 | 3968 | 97 | ||
| XM_009293362.3 | XP_009291637.1 | 17 - 18 | 3180 | 97 | ||
| XM_009293365.3 | XP_009291640.1 | 18 - 19 | 3269 | 97 | ||
| GenBank mRNA | BC128874.1 | - | 9 - 10 | 2187 | 97 | |
More Information
Set Membership: |
|

kdm2aa
Genomic Map