Assay Details
| Gene Symbol: surf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080173.1 | NP_001073642.1 | 8 - 9 | 891 | 75 | |
| XM_009301509.2 | XP_009299784.1 | 5 - 6 | 595 | 75 | ||
| GenBank mRNA | BC129348.1 | - | 8 - 9 | 891 | 75 | |
| BC164262.1 | - | 8 - 9 | 895 | 75 | ||
More Information
Set Membership: |
|

surf1
Genomic Map