Assay Details
| Gene Symbol: rdh10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080583.1 | NP_001074052.1 | 5 - 6 | 951 | 67 | |
| GenBank mRNA | BC129239.1 | - | 5 - 6 | 951 | 67 | |
More Information
Set Membership: |
|

rdh10a
Genomic Map