Assay Details
| Gene Symbol: sacm1lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080624.2 | NP_001074093.2 | 1 - 2 | 73 | 64 | |
| GenBank mRNA | BC129344.1 | - | 1 - 2 | 73 | 64 | |
| BC165691.1 | - | 1 - 2 | 67 | 64 | ||
More Information
Set Membership: |
|

sacm1lb
Genomic Map