Assay Details
| Gene Symbol: loxl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080626.1 | NP_001074095.1 | 5 - 6 | 1035 | 64 | |
| XM_009301553.3 | XP_009299828.1 | 4 - 5 | 2398 | 64 | ||
| GenBank mRNA | BC129226.1 | - | 5 - 6 | 1035 | 64 | |
| BC155756.1 | - | 5 - 6 | 1066 | 64 | ||
| EF030482.1 | - | 4 - 5 | 938 | 64 | ||
More Information
Set Membership: |
|

loxl2b
Genomic Map