Assay Details
| Gene Symbol: loxl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080626.1 | NP_001074095.1 | 12 - 13 | 2207 | 71 | |
| XM_009301553.3 | XP_009299828.1 | 11 - 12 | 3570 | 71 | ||
| GenBank mRNA | BC129226.1 | - | 12 - 13 | 2207 | 71 | |
| BC155756.1 | - | 12 - 13 | 2238 | 71 | ||
| EF030482.1 | - | 11 - 12 | 2110 | 71 | ||
More Information
Set Membership: |
|

loxl2b
Genomic Map