Assay Details
| Gene Symbol: zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082999.1 | NP_001076468.1 | 1293 | 70 | ||
| GenBank mRNA | BC133104.1 | - | 1293 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zbtb49| Gene Symbol: zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082999.1 | NP_001076468.1 | 1293 | 70 | ||
| GenBank mRNA | BC133104.1 | - | 1293 | 70 | ||
Set Membership: |
|