Assay Details
| Gene Symbol: acap3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083015.1 | NP_001076484.1 | 7 - 8 | 715 | 79 | |
| XM_021479534.1 | XP_021335209.1 | 7 - 8 | 733 | 79 | ||
| XM_021479535.1 | XP_021335210.1 | 7 - 8 | 732 | 79 | ||
| GenBank mRNA | BC133093.1 | - | 7 - 8 | 715 | 79 | |
More Information
Set Membership: |
|

acap3b
Genomic Map