Assay Details
| Gene Symbol: acap3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083015.1 | NP_001076484.1 | 12 - 13 | 1062 | 72 | |
| XM_017358180.2 | XP_017213669.1 | 3 - 4 | 534 | 72 | ||
| XM_021479534.1 | XP_021335209.1 | 12 - 13 | 1080 | 72 | ||
| XM_021479535.1 | XP_021335210.1 | 12 - 13 | 1079 | 72 | ||
| GenBank mRNA | BC133093.1 | - | 12 - 13 | 1062 | 72 | |
More Information
Set Membership: |
|

acap3b
Genomic Map