Assay Details
| Gene Symbol: lto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083552.1 | NP_001077021.1 | 2 - 3 | 211 | 67 | |
| GenBank mRNA | BC133920.1 | - | 2 - 3 | 211 | 67 | |
More Information
Set Membership: |
|

lto1
Genomic Map