Assay Details
| Gene Symbol: errfi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083570.1 | NP_001077039.1 | 2 - 3 | 326 | 108 | |
| GenBank mRNA | BC133880.1 | - | 2 - 3 | 326 | 108 | |
More Information
Set Membership: |
|

errfi1a
Genomic Map