Assay Details
| Gene Symbol: psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083573.1 | NP_001077042.1 | 9 - 10 | 831 | 64 | |
| GenBank mRNA | BC133929.1 | - | 9 - 10 | 831 | 64 | |
More Information
Set Membership: |
|

psmd14
Genomic Map