Assay Details
| Gene Symbol: anpepb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089325.1 | NP_001082794.1 | 7 - 8 | 1451 | 92 | |
| GenBank mRNA | BC115260.1 | - | 7 - 8 | 1451 | 92 | |
| BC154244.1 | - | 7 - 8 | 1452 | 92 | ||
More Information
Set Membership: |
|

anpepb
Genomic Map