Assay Details
| Gene Symbol: loxl3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145684.2 | NP_001139156.1 | 13 - 14 | 2420 | 91 | |
| XM_005156544.4 | XP_005156601.1 | 15 - 16 | 2731 | 91 | ||
| XM_009307148.3 | XP_009305423.1 | 14 - 15 | 2561 | 91 | ||
| XM_009307149.3 | XP_009305424.1 | 14 - 15 | 2556 | 91 | ||
| XM_009307150.3 | XP_009305425.1 | 12 - 13 | 2264 | 91 | ||
| XM_017358707.2 | XP_017214196.1 | 15 - 16 | 2733 | 91 | ||
| XM_021480538.1 | XP_021336213.1 | 14 - 15 | 2676 | 91 | ||
| GenBank mRNA | BC139677.1 | - | 3 - 4 | 547 | 91 | |
| EF030484.1 | - | 4 - 5 | 620 | 91 | ||
More Information
Set Membership: |
|

loxl3b
Genomic Map