Assay Details
| Gene Symbol: igf2bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089374.1 | NP_001082843.1 | 10 - 11 | 1335 | 98 | |
| XM_005159948.4 | XP_005160005.1 | 11 - 12 | 1447 | 98 | ||
| XM_005159949.4 | XP_005160006.1 | 11 - 12 | 1611 | 98 | ||
| XM_017354915.2 | XP_017210404.1 | 11 - 12 | 1680 | 98 | ||
| XM_021474969.1 | XP_021330644.1 | 11 - 12 | 1609 | 98 | ||
| GenBank mRNA | BC134152.1 | - | 8 - 9 | 810 | 98 | |
| EU296399.1 | - | 12 - 13 | 1566 | 98 | ||
| EU296400.1 | - | 11 - 12 | 1381 | 98 | ||
More Information
Set Membership: |
|

igf2bp2b
Genomic Map