Assay Details
| Gene Symbol: igf2bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089374.1 | NP_001082843.1 | 2 - 3 | 444 | 91 | |
| XM_005159948.4 | XP_005160005.1 | 3 - 4 | 556 | 91 | ||
| XM_005159949.4 | XP_005160006.1 | 3 - 4 | 720 | 91 | ||
| XM_017354915.2 | XP_017210404.1 | 3 - 4 | 789 | 91 | ||
| XM_021474969.1 | XP_021330644.1 | 3 - 4 | 721 | 91 | ||
| GenBank mRNA | EU296399.1 | - | 4 - 5 | 675 | 91 | |
| EU296400.1 | - | 3 - 4 | 490 | 91 | ||
More Information
Set Membership: |
|

igf2bp2b
Genomic Map