Assay Details
| Gene Symbol: igf2bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089374.1 | NP_001082843.1 | 3 - 4 | 564 | 102 | |
| GenBank mRNA | EU296399.1 | - | 5 - 6 | 795 | 102 | |
| EU296400.1 | - | 4 - 5 | 610 | 102 | ||
More Information
Set Membership: |
|

igf2bp2b
Genomic Map