Assay Details
| Gene Symbol: igf2bp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089374.1 | NP_001082843.1 | 9 - 10 | 1220 | 68 | |
| XM_005159948.4 | XP_005160005.1 | 10 - 11 | 68 | |||
| XM_005159949.4 | XP_005160006.1 | 10 - 11 | 68 | |||
| XM_017354915.2 | XP_017210404.1 | 10 - 11 | 68 | |||
| XM_021474969.1 | XP_021330644.1 | 10 - 11 | 68 | |||
| GenBank mRNA | BC134152.1 | - | 7 - 8 | 68 | ||
| EU296399.1 | - | 11 - 12 | 1451 | 68 | ||
| EU296400.1 | - | 10 - 11 | 1266 | 68 | ||
More Information
Set Membership: |
|

igf2bp2b
Genomic Map