Assay Details
| Gene Symbol: larp6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089408.1 | NP_001082877.1 | 3 - 4 | 364 | 84 | |
| GenBank mRNA | BC134173.1 | - | 3 - 4 | 364 | 84 | |
More Information
Set Membership: |
|

larp6b
Genomic Map