Assay Details
Gene Symbol: zgc:163073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089511.1 | NP_001082980.1 | 5 - 6 | 666 | 100 | |
GenBank mRNA | BC124826.1 | - | 5 - 6 | 611 | 100 | |
BC135098.1 | - | 5 - 6 | 666 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
