Assay Details
| Gene Symbol: cyp4f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089541.1 | NP_001083010.1 | 11 - 12 | 1496 | 62 | |
| GenBank mRNA | BC134144.1 | - | 11 - 12 | 1496 | 62 | |
More Information
Set Membership: |
|

cyp4f3
Genomic Map