Assay Details
| Gene Symbol: cop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089542.1 | NP_001083011.1 | 7 - 8 | 1032 | 149 | |
| XM_021478383.1 | XP_021334058.1 | 7 - 8 | 1109 | 149 | ||
| GenBank mRNA | BC139708.1 | - | 7 - 8 | 1032 | 149 | |
More Information
Set Membership: |
|

cop1
Genomic Map