Assay Details
| Gene Symbol: gad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089570.2 | NP_001083039.2 | 5 - 6 | 654 | 73 | |
| GenBank mRNA | BC135107.1 | - | 4 - 5 | 502 | 73 | |
More Information
Set Membership: |
|

gad3
Genomic Map