Assay Details
| Gene Symbol: gad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089570.2 | NP_001083039.2 | 8 - 9 | 908 | 60 | |
| GenBank mRNA | BC135107.1 | - | 7 - 8 | 756 | 60 | |
More Information
Set Membership: |
|

gad3
Genomic Map