Assay Details
| Gene Symbol: sat1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093748.2 | NP_001087217.1 | 4 - 5 | 479 | 102 | |
| GenBank mRNA | BC150358.1 | - | 4 - 5 | 479 | 102 | |
| BC162066.1 | - | 4 - 5 | 436 | 102 | ||
| BC162087.1 | - | 4 - 5 | 436 | 102 | ||
More Information
Set Membership: |
|

sat1b
Genomic Map