Assay Details
| Gene Symbol: sat1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093748.2 | NP_001087217.1 | 5 - 6 | 521 | 128 | |
| GenBank mRNA | BC150358.1 | - | 5 - 6 | 521 | 128 | |
| BC162066.1 | - | 5 - 6 | 478 | 128 | ||
| BC162087.1 | - | 5 - 6 | 478 | 128 | ||
More Information
Set Membership: |
|

sat1b
Genomic Map