Assay Details
| Gene Symbol: cpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110021.2 | NP_001103491.2 | 10 - 11 | 1061 | 93 | |
| NM_001328425.1 | NP_001315354.1 | 10 - 11 | 1126 | 93 | ||
| GenBank mRNA | BC067637.1 | - | 10 - 11 | 1063 | 93 | |
| BC154043.1 | - | 10 - 11 | 1021 | 93 | ||
More Information
Set Membership: |
|

cpb1
Genomic Map