Assay Details
| Gene Symbol: ppp4r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110371.1 | NP_001103841.1 | 8 - 9 | 1050 | 93 | |
| GenBank mRNA | BC096878.1 | - | 8 - 9 | 1024 | 93 | |
| BC129134.1 | - | 8 - 9 | 1034 | 93 | ||
| BC154497.1 | - | 8 - 9 | 1050 | 93 | ||
More Information
Set Membership: |
|

ppp4r2b
Genomic Map