Assay Details
| Gene Symbol: igsf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110473.1 | NP_001103943.1 | 2 - 3 | 362 | 94 | |
| XM_017353589.2 | XP_017209078.1 | 1 - 2 | 113 | 94 | ||
| GenBank mRNA | BC134916.1 | - | 2 - 3 | 247 | 94 | |
| CU458945.1 | - | 2 - 3 | 392 | 94 | ||
| KM655674.1 | - | 2 - 3 | 330 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017353589.2 transcripts.
Set Membership: |
|

igsf21b
Genomic Map